Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: CYTIIERNNR QMLPVIRSTA GGDSVQICTN PLDTFFPFDP CVLKRSKKFI DPIYQVWEDM SAEELQEFKK PMKKDIVEDE DDDFLKGEVP QNDTVIGITP SSFDTHF
Required for efficient transcription initiation by RNA polymerase I. Required for the formation of the competent preinitiation complex (PIC). Dissociates from pol I as a consequence of transcription. In vitro, cannot activate transcription in a subsequent transcription reaction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RNA polymerase I-specific transcription initiation factor RRN3; RRN3 RNA polymerase I transcription factor homolog; TIF-IA; Transcription initiation factor IA; transcription initiation factor TIF-IA
Gene Aliases: A-270G1.2; RRN3; TIFIA
UniProt ID: (Human) Q9NYV6
Entrez Gene ID: (Human) 54700
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support