Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MVARRRKCA ARDPEDRIPS PLGYAAIPIK FSEKQQASHY LYVRAHGVRQ GTKSTWPQKR TLFVLNVPPY CTEESLSRLL STCGLVQSVE LQEKPDLAES PKESRSKFFH PKPVPGFQVA YVVFQKPSGV SAALALKGPL LVSTESHPVK SGIHKWISDY ADSVPDPEAL RVEVDTF (1-176 aa encoded by BC121118)
RRP7A (Ribosomal RNA Processing 7 Homolog A) is a Protein Coding gene. Diseases associated with RRP7A include Autosomal Dominant Non-Syndromic Intellectual Disability 1 and Autosomal Dominant Non-Syndromic Intellectual Disability. Among its related pathways are rRNA processing in the nucleus and cytosol and Gene Expression. Gene Ontology (GO) annotations related to this gene include nucleotide binding. [GeneCards]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CTA-126B4.5; Gastric cancer antigen Zg14; Gastric cancer antigen Zg14 homolog; kidney highly expressed protein 1; Ribosomal RNA-processing protein 7 homolog A
Gene Aliases: 1110014J01Rik; AA408146; BK126B4.3; CGI-96; Kheg1; RRP7A
UniProt ID: (Human) Q9Y3A4, (Mouse) Q9D1C9
Entrez Gene ID: (Human) 27341, (Mouse) 74778
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support