Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Viperin is a 43kDa protein which is one of several hundred interferon inducible viral proteins. Elevated expression of viperin is induced by LPS and poly I:C through TLR dependent signaling pathways. Sendai virus mediated viperin expression is induced independently of TLR signaling pathways through RLRs - RIG-I and MAVs (Severa, 2006). Viperin has been shown to play an important role in interferon mediated anti-viral activity in a Hepatitis C virus model system (Helbig, 2011). Viperin is induced in lymphoid cells and dendritic cells (DCs) and highly induced in neutrophils and macrophages during acute LCMV infection in mice (Hinson, ). Viperin functions as a mediator of viral resistance and as a marker for interferon responsiveness making its detection and mechanism of action through specific antibody probes valuable for further studies.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytomegalovirus-induced gene 5 protein; Radical S-adenosyl methionine domain-containing protein 2; RSAD2; S-adenosylmethionine-dependent nucleotide dehydratase RSAD2; SAND; Viperin; Virus inhibitory protein, endoplasmic reticulum-associated, interferon-inducible
Gene Aliases: 2510004L01Rik; cig33; CIG5; RSAD2; vig1
UniProt ID: (Human) Q8WXG1
Entrez Gene ID: (Human) 91543
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support