Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GRRQRRMHP NVSQGCQGGC ATCSDYNGCL SCKPRLFFAL ERIGMKQIGV CLSSCPSGYY GTRYPDINKC TKCKADCDTC FNKNFCTKCK SGFYLHLGKC LDNCPEGLEA NNHTMECVSI VHCEVSEWNP WSPCTKKGKT CGFKRGTETR VREIIQHPSA KGNLCPPTNE TRKCTVQRKK CQKGERGKKG RERKRKKPNK GESKEAIPDS KSLESSKEIP EQRENKQQQK KRKVQDKQKS VSVSTVH (27-272 aa encoded by BC022367)
RSPO3 is a member of the R-spondin family and plays a role in the regulation of Wnt (wingless-type MMTV integration site family)/beta-catenin and Wnt/planar cell polarity (PCP) signaling pathways, which are involved in development, cell growth and disease pathogenesis. RSPO3 has been shown to be involved in regulating the balance between angioblast and blood cell specification during embryonic vasculogenesis and angiogenesis and may be involved in tumor development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPWTSR; hRspo3; Protein with TSP type-1 repeat; R-spondin 3 homolog; R-spondin-3; Roof plate-specific spondin-3; Thrombospondin type-1 domain-containing protein 2; thrombospondin, type I, domain containing 2
Gene Aliases: CRISTIN1; PWTSR; RSPO3; THSD2
UniProt ID: (Human) Q9BXY4
Entrez Gene ID: (Human) 84870
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support