Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute at 0.2 mg/mL in sterile PBS.
Endoxin level is <0.10 EU per 1 µg of the antibody by the LAL method.
RSPO4 is a member of the R-spondin family of proteins that share a common domain organization consisting of a signal peptide, cysteine-rich/furin-like domain, thrombospondin domain and a C-terminal basic region. The encoded protein may be involved in activation of Wnt/beta-catenin signaling pathways. Mutations in this gene are associated with anonychia congenital. Alternate splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cristin-4; Cysteine-rich and single thrombospondin domain-containing protein 4; mCristin-4; R-spondin-4; R-spondin-like protein; Roof plate-specific spondin-4
Gene Aliases: A730099F22; A930029K19Rik; Rspo4
UniProt ID: (Mouse) Q8BJ73
Entrez Gene ID: (Mouse) 228770
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support