Search Thermo Fisher Scientific
FIGURE: 1 / 3
This product can be used in Sandwich ELISA to detect RSV NP using MA5-46972 as a capture antibody and MA5-50309 as the detection antibody.
Protein Aliases: human respiratory syncytial virus; RSV; RSV NP; RSV Nuclear Protein