Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKKQANKTA SSGSSDKDSS AESSAPEEGE VSDSDSNSSS SSSDSDSSSE DEEFHDGYGE DLMGDEEDRA RLEQMTEKER EQELFNRIEK REVLKRRFEI KKKLKTAKKK EKKEKKKKQE EEQEKKKLTQ IQESQVTSHN KERRSKRDEK LDKKSQAMEE LKAEREKRKN RTAELLAKKQ PLKTSEVYSD DEEEEEDDKS SEKSDRSSRT SSSDEEEEKE EIPPKSQPVS LPEELNRVRL SRHKLERWCH MPFFAKTVTG CFVRIGIGNH NSKPVYRVAE ITGVVETAKV YQLGGTRTNK GLQLRHGN (1-307 aa encoded by BC015052)
Three isoforms of serine/threonine kinases, designated alphaPAK p68, betaPAK p65 and gammaPAK p62, have been shown to exhibit a high degree of sequence homology with the S. cerevisiae kinase Ste 20, involved in pheromone signaling. The alpha, beta and gammaPAK isoforms complex specifically with Rac1 and Cdc42 in their active GTP-bound state, inhibiting their intrinsic GTPase activity leading to their autophosphorylation. There are eight sites of autophosphorylation on gammaPAK, including Ser 19, Ser 141 and Thr 402, and phosphorylation of Ser 141 and Thr 402 is correlated with gammaPAK activation. Once phosphorylated and their affinity for Rac/Cdc42 reduced, the PAK isoforms disassociate from the complex to seek downstream substrates.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gene trap locus 7; ortholog of mouse gene trap locus 7; RNA polymerase-associated protein RTF1 homolog; Rtf1 Paf1/RNA polymerase II complex component homolog; Rtf1, Paf1/RNA polymerase II complex component, homolog
Gene Aliases: 2900005O08Rik; 6530416A09Rik; AI853581; AU043144; AW553985; cbp82; GTL7; KIAA0252; RTF1
UniProt ID: (Human) Q92541, (Mouse) A2AQ19
Entrez Gene ID: (Human) 23168, (Rat) 366169, (Mouse) 76246
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support