Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MASNSIFDSF PTYSPTFIRD PSTSRRFTPP SPAFPCGGGG GKMGENSGAL SAQAAVGPGG RARPEVRSMV DVLADHAGEL VRTDSPNFLC SVLPSHWRCN KTLPVAFKVV ALGDVPDGTV VTVMAGNDEN YSAELRNASA VMKNQVARFN DLRFVGRSGR GKSFTLTITV FTNPTQVATY HRAIKVTVDG PREPRRHRQK LEDQTKPFPD RFGDLERLRM RVTPSTPSPR GSLSTTSHFS SQPQTPIQGT SELNPFSDPR QFDRSFPTLP TLTESRFPDP RMHYPGAMSA AFPYSATPSG TSISSLSVAG MPATSRFHHT YLPPPYPGAP QNQSGPFQAN PSPYHLYYGT SSGSYQFSMV AGSSSGGDRS PTRMLASCTS SAASVAAGNL MNPSLGGQSD GVEADGSHSN SPTALSTPGR MDEAVWRPY
This gene encodes a member of the runt domain-containing family of transcription factors. A heterodimer of this protein and a beta subunit forms a complex that binds to the core DNA sequence 5'-PYGPYGGT-3' found in a number of enhancers and promoters, and can either activate or suppress transcription. It also interacts with other transcription factors. It functions as a tumor suppressor, and the gene is frequently deleted or transcriptionally silenced in cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acute myeloid leukemia 2 protein; acute myeloid leukemia gene 2; CBF-alpha 3; CBF-alpha-3; Core-binding factor subunit alpha-3; core-binding factor, runt domain, alpha subunit 3; Oncogene AML-2; PEA2 alpha C; PEA2-alpha C; PEBP2 alpha C; PEBP2-alpha C; Polyomavirus enhancer-binding protein 2 alpha C subunit; Runt-related transcription factor 3; SL3-3 enhancer factor 1 alpha C subunit; SL3/AKV core-binding factor alpha C subunit; transcription factor AML2
Gene Aliases: AML2; CBFA3; PEBP2A3; PEBP2aC; RUNX3
UniProt ID: (Human) Q13761
Entrez Gene ID: (Human) 864
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support