Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a RUN and SH3 domain containing protein that interacts with Rab1b and Rab1-binding protein GM130. Alternatively spliced transcript variants have been observed for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AP-4 complex accessory subunit RUSC2; Interacting protein of Rab1; Iporin; RUN and SH3 domain-containing protein 2
Gene Aliases: AI840675; Iporin; KIAA0375; mKIAA0375; RUSC2
UniProt ID: (Human) Q8N2Y8
Entrez Gene ID: (Human) 9853, (Mouse) 100213
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support