Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
FIGURE: 1 / 2
Rab proteins belong to the Ras small GTPase superfamily. The GTP-binding Rab3 subfamily has four known isoforms: 3A, 3B, 3C and 3D. These proteins are expressed in the highest concentration in the brain and endocrine tissues and are involved in regulated exocytosis of hormones and neurotransmitters. Specifically Rab3's are thought to play a role in presynaptic vesicle trafficking and priming.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brain antigen RAB3B; glioblastoma overexpressed; GTP-binding protein Rab-3D; Rab3D upregulated with myeloid differentiation; RAS-associated protein RAB3A; Ras-related protein Rab-3A; Ras-related protein Rab-3B; Ras-related protein Rab-3C; Ras-related protein Rab-3D; Ras-related small GTP binding protein 3A; small GTP-binding protein Rab3C
Gene Aliases: 2610528C18Rik; 2700062I01Rik; 3110015B08Rik; 3110037E15Rik; AI850886; C130057E11Rik; D2-2; GOV; RAB16; RAB3; RAB3A; RAB3B; RAB3C; RAB3D; RAD3D
UniProt ID: (Human) P20336, (Human) P20337, (Human) Q96E17, (Human) O95716, (Rat) P63012, (Mouse) P63011, (Mouse) Q9CZT8, (Rat) Q63941, (Rat) P62824, (Mouse) P62823, (Mouse) P35276, (Rat) Q63942
Entrez Gene ID: (Human) 5864, (Human) 5865, (Human) 115827, (Human) 9545, (Rat) 25531, (Mouse) 19339, (Mouse) 69908, (Rat) 81755, (Rat) 171058, (Mouse) 67295, (Mouse) 19340, (Rat) 140665
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support