Invitrogen
FIGURE: 1 / 6
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration.
Product # 31194 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications.
Product # 31194 reacts with the heavy chains of mouse IgG but not with the light chains of most mouse immunoglobulins. This antibody does not react against mouse IgM or non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species.
Store product at 4°C until opened. After opening, dilute only enough antibody for a single day’s use. Store remainder at 4°C under sterile conditions.
Country of Origin: USA
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.