Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration.
Product # 31627 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications.
Product # 31627 reacts with whole molecule sheep IgG. It also reacts with the light chains of other sheep immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species.
Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C.
Fluorophore: Fluorescein-5-isothiocyanate (FITC-isomer 1)
Amax = 492 nm; Emax = 520 nm
Fluorophore/Protein: 9.1 µg/mg; 3.4 moles FITC per mole IgG
Reconstitute with 1.1 mL of distilled water (1.5 mg/mL after restoration).
Country of Origin: USA
Anti-Sheep secondary antibodies are affinity-purified antibodies with well-characterized specificity for ovine (sheep) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support