Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: DSIIVFYNSV FMQRLKTNIL QYASTRPPTL SPIPHIPRSP YKFPSSPLRI PGGNIYISPL KSPYKISEGL PTPTKMTPRS RILVSIGESF GTSEKFQKIN Q
Rb is a tumor suppressor nuclear phosphoprotein capable of binding to DNA. It is phosphorylated on serine and threonine, but not on tyrosine residues. It forms a complex with SV40 large T antigen, adenovirus E1A, and human papilloma virus-16 E. Rb protein may act by regulating transcription and loss of its function leads to uncontrolled cell growth. Aberrations in the RB gene have been implicated in cancers of breast, colon, prostate, kidney, nasopharynx, and leukemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cleaved Rb; delta RB-p70; DRb-p70; P105RB; p110-RB1; Phospho-Retinoblastoma; pp105; pRb; Rb; Rb-p70; Retin; Retinoblastoma; Retinoblastoma 1 (including osteosarcoma); retinoblastoma gene; Retinoblastoma-associated protein
Gene Aliases: pRb; Rb; Rb-1; Rb1
UniProt ID: (Rat) P33568, (Mouse) P13405
Entrez Gene ID: (Rat) 24708, (Mouse) 19645
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support