Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TFCEPYGPRE LITTGLNTTT ASVLQFSIGS GSCRFSYSDP SIIVLYAKNN SADWIQLEKI RAPSNVSTII HILYLPEDAK GENVQFQWKQ ENLRVGEVYE
Reelin in an extracellular matrix serine protease that plays a role in layering of neurons in the cerebral cortex and cerebellum. Regulates microtubule function in neurons and neuronal migration. Affects migration of sympathetic preganglionic neurons in the spinal cord, where it seems to act as a barrier to neuronal migration. Enzymatic activity is important for the modulation of cell adhesion. Binding to the extracellular domains of lipoprotein receptors VLDLR and LRP8/APOER2 induces tyrosine phosphorylation of DAB1 and modulation of TAU phosphorylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: extracellular matrix serine protease; Reeler protein; Reelin
Gene Aliases: ETL7; LIS2; PRO1598; Reelen; reeler; RELN; RL
UniProt ID: (Human) P78509, (Mouse) Q60841, (Rat) P58751
Entrez Gene ID: (Human) 5649, (Mouse) 19699, (Rat) 24718
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support