Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
RHOBTB3 Polyclonal Antibody detects endogenous levels of RHOBTB3 protein.
The Rho subfamily of Ras-related GTPases controls multiple aspects of cell function, including cytoskeletal rearrangement, nuclear signaling and cell growth. RHOBTB1 (Rho-related BTB domain-containing protein 1) and RHOBTB3 (Rho-related BTB domain-containing protein 3) each contain two BTB (POZ) domains and belong to the RhoBTB subfamily of Rho GTPases. Members of the RhoBTB subfamily are evolutionarily conserved and are characterized by a proline-rich region, a GTPase domain and two tandem BTB repeats. While both RHOBTB1 and RHOBTB3 are expressed ubiquitously, RHOBTB1 is found at high levels in placenta, stomach, testis, kidney and skeletal muscle, whereas RHOBTB3 is found at high levels in neural and cardiac tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Rho-related BTB domain containing 3; Rho-related BTB domain-containing protein 3; RHOBTB 3
Gene Aliases: 1700040C17Rik; 2610033K01Rik; 4930503C18Rik; AI317148; AW208826; KIAA0878; mKIAA0878; RHOBTB3
UniProt ID: (Human) O94955, (Mouse) Q9CTN4
Entrez Gene ID: (Human) 22836, (Mouse) 73296
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support