Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Control: 293T, LOVO, SH-SY-5Y, human fetal skeletal muscle tissue, mouse brain tissue, mouse cerebellum tissue, rat skeletal muscle tissue.
The ryanodine receptor (RyR) is the channel responsible for the release of calcium from the sarcoplasmic reticulum (SR) in muscle cells and also plays a role in calcium regulation in non-muscle cells. The RyR exists as a homotetramer and is predicted to have a short cytoplasmic C-terminus and 4-10 transmembrane domains; the remainder of the protein, termed the "foot" region is located in the cytoplasm between the T-tubule and the SR. The mammalian RyR is the product of three different genes: RyR-1, which is expressed predominantly in skeletal muscle and areas of the brain, RyR-2, which is expressed predominantly in the heart muscle but also found in the stomach, endothelial cells and diffuse areas of the brain, and RyR-3 which is found in smooth muscle and the brain (striatum, thalamus and hippocampus).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cardiac muscle ryanodine receptor; Cardiac muscle ryanodine receptor-calcium release channel; cardiac-type ryanodine receptor; central core disease of muscle; islet-type ryanodine receptor; kidney-type ryanodine receptor; protein phosphatase 1, regulatory subunit 137; Ryanodine receptor 1; ryanodine receptor 1 (skeletal); Ryanodine receptor 2; ryanodine receptor 2 (cardiac); RYR-1; RYR-2; sarcoplasmic reticulum calcium release channel; Skeletal muscle calcium release channel; Skeletal muscle ryanodine receptor; Skeletal muscle-type ryanodine receptor; Type 1 ryanodine receptor; type 1-like ryanodine receptor; Type 2 ryanodine receptor
Gene Aliases: ARVC2; ARVD2; CCO; MHS; MHS1; PPP1R137; RYDR; RYR; RYR-1; RYR-2; RYR1; RYR2; SKRR; VTSIP
UniProt ID: (Human) P21817, (Human) Q92736
Entrez Gene ID: (Human) 6261, (Human) 6262
Molecular Function:
ligand-gated ion channel
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support