Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
S-antigen, also identified as arrestin, is a protein involved in the regulation of phototransduction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 48 kDa protein; arrestin 1; arrestin-1; DKFZp686D1084; DKFZp686I1383; Retinal S-antigen; retinal S-antigen (48 KDa protein); rod arrestin; Rod photoreceptor arrestin; S-AG; S-antigen; retina and pineal gland (arrestin); S-arrestin; visual arrestin 1
Gene Aliases: A930001K18Rik; Arr1; arrestin; Irbp; RP47; S-AG; SAG
UniProt ID: (Human) P10523, (Mouse) P20443
Entrez Gene ID: (Human) 6295, (Mouse) 20215
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support