Search Thermo Fisher Scientific
This antibody can detect the over-expressed fusion proteins containing S1 epitope tag.
S1 tag, with sequence NANNPDWDF, is derived from the hepatitis B virus preS1 region. S1 tag is widely used in various protein expression systems.
Protein Aliases: hepatitis B virus preS1; NANNPDWDF