Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive control: Melanoma or Schwannoma
Cellular location: Cytoplasmic.
S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein S100-A1; Protein S100-B; S 100; S-100 calcium-binding protein, beta chain; S-100 protein alpha chain; S-100 protein beta chain; S-100 protein subunit alpha; S-100 protein subunit beta; S100 alpha; S100 calcium-binding protein A1; S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S100 calcium-binding protein, beta (neural); S100 protein, alpha polypeptide; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural; S100alpha
Gene Aliases: AI266795; AI850290; Bpb; NEF; S100; S100-alpha; S100-B; S100A; S100A1; S100B; S100beta; S100P
UniProt ID: (Bovine) P02639, (Human) P23297, (Human) P04271, (Bovine) P02638, (Rat) P35467, (Mouse) P56565, (Mouse) P50114, (Rat) P04631
Entrez Gene ID: (Bovine) 528735, (Human) 6271, (Human) 6285, (Bovine) 525716, (Rat) 295214, (Mouse) 20193, (Mouse) 20203, (Rat) 25742
Molecular Function: calmodulin-related
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support