Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAEPLTEL EESIETVVTT FFTFARQEGR KDSLSVNEFK ELVTQQLPHL LKDVGSLDEK MKSLDVNQDS ELKFNEYWRL IGELAKEIRK KKDLKIRKK (1-98 aa encoded by BC000632)
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein is widely expressed in various types of tissues with a high expression level in thyroid gland. In smooth muscle cells, this protein co-expresses with other family members in the nucleus and in stress fibers, suggesting diverse functions in signal transduction. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein S100-A13; S100 calcium-binding protein A13
Gene Aliases: S100A1; S100A13
UniProt ID: (Human) Q99584, (Mouse) P97352
Entrez Gene ID: (Human) 6284, (Rat) 295213, (Mouse) 20196
Molecular Function:
calmodulin-related
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support