Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
Positive Control: Human intestine stomach or kidney.
Cellular Location: Cytoplasm.
S100G encodes calbindin D9K, a vitamin D-dependent calcium-binding protein. This cytosolic protein belongs to a family of calcium-binding proteins that includes calmodulin, parvalbumin, troponin C, and S100 protein. In the intestine, the protein is vitamin D-dependent and its expression correlates with calcium transport activity. The protein may increase Ca2+ absorption by buffering Ca2+ in the cytoplasm and increase ATP-dependent Ca2+ transport in duodenal basolateral membrane vesicles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CABP; calbindin 3, (vitamin D-dependent calcium-binding protein); calbindin D9K; Calbindin-D9k; Protein S100-G; S100 calcium-binding protein G; Vitamin D-dependent calcium-binding protein, intestinal
Gene Aliases: CABP; CABP1; CABP9K; CALB3; S100D; S100G
UniProt ID: (Human) Q5JS49
Entrez Gene ID: (Human) 795
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support