Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kruppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. ANKZF1 (ankyrin repeat and zinc finger domain containing 1), also known as ZNF744, is 726 amino acids in length and lacks a KRAB domain but contains one C2H2-type zinc finger and two ANK repeats. The gene encoding ANKZF1 localizes to chromosome 2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686B15167; DnaJ homolog subfamily C member 29; DNAJC29; protein phosphatase 1, regulatory subunit 138; Sacsin; spastic ataxia of Charlevoix-Saguenay (sacsin)
Gene Aliases: A230052M14; ARSACS; DNAJC29; E130115J16Rik; KIAA0730; mKIAA0730; PPP1R138; SACS; SPAX6
UniProt ID: (Human) Q9NZJ4, (Mouse) Q9JLC8
Entrez Gene ID: (Human) 26278, (Mouse) 50720
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support