Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: NGLEDDSRDG QEDMEASLEN LQNMGMMDMS VLDETEVANS SAPDFGEDGT DGLLDSFCDS KEYVAAQLRQ LPAQPPEHAV DGEGFKNTLE TSSLNFK
Binds to scaffold/matrix attachment region (S/MAR) DNA. Can function as an estrogen receptor corepressor and can also inhibit cell proliferation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SAF-B2; Scaffold attachment factor B2
Gene Aliases: KIAA0138; SAFB2
UniProt ID: (Human) B4DKG3
Entrez Gene ID: (Human) 9667
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support