Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
This gene encodes a protein containing multiple zinc finger domains. The encoded protein functions in optical fissure closure during development of the eye in the embryo. Mutations in this gene are associated with ocular coloboma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Sal-2; sal-like 2; Sal-like protein 2; Spalt-like protein 2; Zinc finger protein 795; Zinc finger protein SALL2; Zinc finger protein Spalt-2
Gene Aliases: AI225809; AW559097; COLB; HSAL2; KIAA0360; mKIAA0360; Msal-2; p150(Sal2); Sal-2; SAL2; SALL2; ZNF795
UniProt ID: (Human) Q9Y467, (Mouse) Q9QX96
Entrez Gene ID: (Human) 6297, (Mouse) 50524, (Rat) 305854
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support