Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: NRGRSDSVDY GQTHYYHQRQ NSDDKLNGWQ NSRDSGICIN ASNWQDKSMG CENGHVPLYS SSSVPTTINT IGTSTSTILS GQAHHSPLKR SVSLTPPMNV PNQPLGHGWM
Sterile alpha motifs (SAMs) in proteins such as SAMD4A are part of an RNA-binding domain that functions as a posttranscriptional regulator by binding to an RNA sequence motif known as the Smaug recognition element, which was named after the Drosophila Smaug protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein Smaug homolog 1; SAM domain-containing protein 4A; Smaug 1; Sterile alpha motif domain-containing protein 4A
Gene Aliases: KIAA1053; SAMD4; SAMD4A; SMAUG; SMAUG1; SMG; SMGA
UniProt ID: (Human) Q9UPU9
Entrez Gene ID: (Human) 23034
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support