Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
Predicted to react in Mouse, Rat
Has transcriptional repressor activity. Overexpression inhibits the transcriptional activities of AP-1, p53/TP53 and CDKN1A. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hSmaug2; Protein Smaug homolog 2; SAM domain-containing protein 4B; Smaug 2; smaug homolog B; Sterile alpha motif domain-containing protein 4B
Gene Aliases: SAMD4B; SMAUG2; SMGB
UniProt ID: (Human) Q5PRF9
Entrez Gene ID: (Human) 55095
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support