Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Involved in the regulation of gene expression in the retina. It functions as a negative regulator of CRX-controlled genes. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SAM domain-containing protein 7; Sterile alpha motif domain-containing protein 7
Gene Aliases: 4930597A01Rik; SAMD7
UniProt ID: (Human) Q7Z3H4, (Mouse) Q8C8Y5
Entrez Gene ID: (Human) 344658, (Mouse) 75953, (Rat) 310257
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support