Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
This gene may play a role in regulation of the innate immune response. The encoded protein is upregulated in response to viral infection and may be involved in mediation of tumor necrosis factor-alpha proinflammatory responses. Mutations in this gene have been associated with Aicardi-Goutieres syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DCIP; Dendritic cell-derived IFNG-induced protein; Deoxynucleoside triphosphate triphosphohydrolase SAMHD1; dNTPase; hSAMHD1; IFN-gamma induced; Interferon-gamma-inducible protein Mg11; Monocyte protein 5; MOP-5; mSAMHD1; RP1-132F21.1; SAM domain and HD domain 1; SAM domain and HD domain-containing protein 1
Gene Aliases: CHBL2; DCIP; E330031J07Rik; HDDC1; Mg11; Mg21; MOP-5; MOP5; SAMHD1; SBBI88
UniProt ID: (Human) Q9Y3Z3, (Mouse) Q60710
Entrez Gene ID: (Human) 25939, (Dog) 485862, (Rat) 311580, (Mouse) 56045
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support