The recommended shelf life for this product is 1 year from date of receipt.
Signal-recognition-particle assembly has a crucial role in targeting secretory proteins to the rough endoplasmic reticulum membrane. SRP9 together with SRP14 and the Alu portion of the SRP RNA, constitutes the elongation arrest domain of SRP. The complex of SRP9 and SRP14 is required for SRP RNA binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 30 kDa Sin3-associated polypeptide; Histone deacetylase complex subunit SAP30; SAP30; sin3 associated polypeptide, 30kD; Sin3 corepressor complex subunit SAP30; Sin3-associated polypeptide p30; sin3-associated polypeptide, 30 kDa; Sin3-associated polypeptide, 30kDa; Sin3-associated protein; Sin3A associated protein 30kDa; Sin3A-associated protein, 30kDa
Gene Aliases: 30kDa; SAP30
UniProt ID: (Human) O75446, (Mouse) O88574
Entrez Gene ID: (Human) 8819, (Mouse) 60406
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support