Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Regulatory subunit of protein phosphatase 6 (PP6). May function as a scaffolding PP6 subunit. Involved in the PP6- mediated dephosphorylation of NFKBIE opposing its degradation in response to TNF-alpha.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: protein phosphatase 6, regulatory subunit 2; SAPS domain family member 2; SAPS domain family, member 2; Serine/threonine-protein phosphatase 6 regulatory subunit 2
Gene Aliases: KIAA0685; PP6R2; PPP6R2; SAP190; SAPS2
UniProt ID: (Human) O75170
Entrez Gene ID: (Human) 9701
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support