Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
The protein encoded by this gene is a small GTPase that acts as a homodimer. The encoded protein is activated by the guanine nucleotide exchange factor PREB and is involved in protein transport from the endoplasmic reticulum to the Golgi. This protein is part of the COPII coat complex. Defects in this gene are a cause of chylomicron retention disease (CMRD), also known as Anderson disease (ANDD). Two transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2310075M17Rik; GTBPB; GTP-binding protein B; GTP-binding protein SAR1b; GTP-binding protein Sara; SAR1 gene homolog B; SAR1 homolog B; SAR1a gene homolog 2; SAR1b gene homolog; secretion associated, Ras related GTPase 1B; Secretion-associated Ras-related GTPase 1B; Small COPII coat GTPase SAR1B; small GTP binding protein
Gene Aliases: 2310075M17Rik; 2900019I22Rik; ANDD; CMRD; GTBPB; SAR1B; SARA1B; SARA2; SARB
UniProt ID: (Human) Q9Y6B6, (Pig) Q5PYH3, (Rat) Q5HZY2, (Mouse) Q9CQC9
Entrez Gene ID: (Rabbit) 100340384, (Human) 51128, (Pig) 494017, (Rat) 287276, (Mouse) 66397
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support