Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Immunogen sequence: MESLVPGFNE KTHVQLSLPV LQVRDVLVRG FGDSVEEVLS EARQHLKDGT CGLVEVEKGV LPQLEQPYVF IKRSDARTAP HGHVMVELVA ELEGIQYGRS GETLGVLVPH VGEIPVAYRK VLLRKNGNKG AGGHSYGADL KSFDLGDELG TDPYEDFQEN WNTKHSSGVT RELMRELNGG AYTRYVDNNF CGPDGYPLEC IKDLLARAGK ASCTLSEQLD FIDTKRGVYC CREHEHEIAW YTERSEKSYE LQTPFEIKLA KKFDTFNGEC PNFVFPLNSI IKTIQPRVEK KKLDGFMGRI RSVYPV
Severe Acute Respiratory Syndrome (SARS), an emerging disease characterized by atypical pneumonia, has been attributed to a novel coronavirus (SARS-CoV). The SARS 3C-like protease (SARS 3CL (pro)) is a cysteine protease engaging in the proteolytic cleavage of the viral precursor polyprotein to a series of functional proteins required for corona virus replication and is considered an attractive target for therapeutics against SARS.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coronavirus; COVID; COVID 19; COVID-19; SARS Coronavirus; SARS CoV; SARS CoV2; SARS-CoV-2
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support