Invitrogen
FIGURE: 1 / 3
Store as concentrated solution.
Centrifuge briefly prior to opening vial.
Responsible for replication and transcription of the viral RNA genome.
Protein Aliases: COVID-19 Nsp12; COVID19 Nsp12; SARS-CoV-2 Nonstructural Protein 12; SARS-COV-2 NSP12; SARS-CoV2 NSP12