FIGURE: 1 / 2
Positive test controls include: 293T.
Immunogen sequence: AVGACVLCNS QTSLRCGACI RRPFLCCKCC YDHVISTSHK LVLSVNPYVC NAPGCDVTDV TQLYLGGMSY YCKSHKPPIS FPLCANGQVF GLYKNTCVGS DNVTDFNAIA TCDWTNAGDY ILANTCTERL KLFAAETLKA TEETFKLSYG IATVREVLSD RELHLSWEVG KPRPPLNRNY VFTGYRVTKN SKVQIGEYTF EKGDYGDAVV YRGTTTYKLN VGDYFVLTSH TVMPLSAPTL VPQEHYVRIT GLYPTLNISD EFSSNVANYQ KVGMQKYSTL QGPPGTGKSH FAIGLALYYP SARIVYTACS HAAVDALCEK ALKYLPIDKC SRIIPARARV ECFDKFKVNS TLEQYVFCTV NALPETTADI VVFDEISMAT NYDLSVVNAR LRAKHYVYIG DPAQLPAPRT LLTKGTLEPE YFNSVCRLMK TIGPDMFLGT CRRCPAEIVD TVSALVYDNK LKAHKDKSAQ CFKMFYKGVI THDVSSAINR PQIGVVREFL TRNPAWRKAV FISPYNSQNA VASKILGLPT QTVDSSQGSE YDYVIFTQTT ETAHSCNVNR FNVAITRAKV GILCIMSDRD LYDKLQFTSL EIPRRNVATL Q
The coronavirus nonstructural protein 13 (nsp13) is one of the SARS-Coronavirus replicase cleaving products encoded by ORF1b. Nsp13 is thought to be part of the viral replication complex, which is associated with intracellular membranes. Nsp13 contains a C-terminal NTPase/Helicase domain and an N-terminal putative zinc-binding motif.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: COVID-19 Envelope; SARS-CoV-2; SARS-CoV-2 Envelope
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support