Product References
SARS-CoV-2 protein ORF8 limits expression levels of Spike antigen and facilitates immune evasion of infected host cells.
The Journal of biological chemistry
Kim IJ,Lee YH,Khalid MM,Chen IP,Zhang Y,Ott M,Verdin E
Published figure using SARS/SARS-CoV-2 Spike Protein S2 monoclonal antibody (Product # MA5-35946) in Immunocytochemistry
Tue Aug 01 00:00:00 EDT 2023
Design and preclinical evaluation of a universal SARS-CoV-2 mRNA vaccine.
Frontiers in immunology
Qin J,Jeon JH,Xu J,Langston LK,Marasini R,Mou S,Montoya B,Melo-Silva CR,Jeon HJ,Zhu T,Sigal LJ,Xu R,Zhu H
MA5-35946 was used in Western Blotting to demonstrate that induction of T cell responses, in the absence of preexisting antibodies, is sufficient to confer protection against severe disease, and that a vaccine containing mRNAs encoding both the Spike and MTE could be further developed as a universal SARS-CoV-2 vaccine.
Tue Apr 11 00:00:00 EDT 2023
Mutations in SARS-CoV-2 variants of concern link to increased spike cleavage and virus transmission.
Cell host & microbe
Escalera A,Gonzalez-Reiche AS,Aslam S,Mena I,Laporte M,Pearl RL,Fossati A,Rathnasinghe R,Alshammary H,van de Guchte A,Farrugia K,Qin Y,Bouhaddou M,Kehrer T,Zuliani-Alvarez L,Meekins DA,Balaraman V,McDowell C,Richt JA,Bajic G,Sordillo EM,Dejosez M,Zwaka TP,Krogan NJ,Simon V,Albrecht RA,van Bakel H,García-Sastre A,Aydillo T
MA5-35946 was used in Western Blot to characterize emerging SARS-CoV-2 spike polymorphisms in vitro and in vivo to understand their impact on transmissibility and virus pathogenicity and fitness.
Wed Mar 09 00:00:00 EST 2022
Zonulin Antagonist, Larazotide (AT1001), As an Adjuvant Treatment for Multisystem Inflammatory Syndrome in Children: A Case Series.
Critical care explorations
Yonker LM,Swank Z,Gilboa T,Senussi Y,Kenyon V,Papadakis L,Boribong BP,Carroll RW,Walt DR,Fasano A
MA5-35946 was used in Neutralization to suggest that expansion of clinical trials is urgently needed to ascertain the clinical impact of larazotide on MIS-C.
Tue Feb 01 00:00:00 EST 2022
Development of a hybrid alphavirus-SARS-CoV-2 pseudovirion for rapid quantification of neutralization antibodies and antiviral drugs.
Cell reports methods
Hetrick B,Chilin LD,He S,Dabbagh D,Alem F,Narayanan A,Luchini A,Li T,Liu X,Copeland J,Pak A,Cunningham T,Liotta L,Petricoin EF,Andalibi A,Wu Y
MA5-35946 was used in Miscellaneous, ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot to demonstrate that Ha-CoV-2 can be used as a robust platform for the rapid quantification of neutralizing antibodies against SARS-CoV-2 and its emerging variants.
Mon Mar 28 00:00:00 EDT 2022
Protocol to isolate and assess spike protein cleavage in SARS-CoV-2 variants obtained from clinical COVID-19 samples.
STAR protocols
Escalera A,García-Sastre A,Aydillo T
MA5-35946 was used in Western Blotting to present a comprehensive protocol to assess SARS-CoV-2 spike protein cleavage in viral supernatants from SARS-CoV-2-infected cells.
Fri Sep 16 00:00:00 EDT 2022
Multiple sites on SARS-CoV-2 spike protein are susceptible to proteolysis by cathepsins B, K, L, S, and V.
Protein science : a publication of the Protein Society
Bollavaram K,Leeman TH,Lee MW,Kulkarni A,Upshaw SG,Yang J,Song H,Platt MO
MA5-35946 was used in Western Blot to highlight cathepsins B, K, L, S, and V for consideration in SARS-CoV-2 infection and presents methodologies by which other proteases can be screened to determine a role in viral entry.
Tue Jun 01 00:00:00 EDT 2021
Immunogenicity and efficacy of mRNA COVID-19 vaccine MRT5500 in preclinical animal models.
NPJ vaccines
Kalnin KV,Plitnik T,Kishko M,Zhang J,Zhang D,Beauvais A,Anosova NG,Tibbitts T,DiNapoli J,Ulinski G,Piepenhagen P,Cummings SM,Bangari DS,Ryan S,Huang PD,Huleatt J,Vincent D,Fries K,Karve S,Goldman R,Gopani H,Dias A,Tran K,Zacharia M,Gu X,Boeglin L,Abysalh J,Vargas J,Beaulieu A,Shah M,Jeannotte T,Gillis K,Chivukula S,Swearingen R,Landolfi V,Fu TM,DeRosa F,Casimiro D
Published figure using SARS/SARS-CoV-2 Spike Protein S2 monoclonal antibody (Product # MA5-35946) in Immunocytochemistry
Mon Apr 19 00:00:00 EDT 2021
SARS-CoV-2 Spike Targets USP33-IRF9 Axis via Exosomal miR-148a to Activate Human Microglia.
Frontiers in immunology
Mishra R,Banerjea AC
MA5-35946 was used in Western Blot to provide novel and relevant insights regarding the impact of Spike gene on shuttling of host microRNAs via exosomes to trigger the neuroinflammation.
Fri May 14 00:00:00 EDT 2021
SARS-CoV-2 Spike Targets USP33-IRF9 Axis via Exosomal miR-148a to Activate Human Microglia.
Frontiers in immunology
Mishra R,Banerjea AC
MA5-35946 was used in Western Blot to provide novel and relevant insights regarding the impact of Spike gene on shuttling of host microRNAs via exosomes to trigger the neuroinflammation.
Fri May 14 00:00:00 EDT 2021
SARS-CoV-2 colonization of maternal and fetal cells of the human placenta promotes alteration of local renin-angiotensin system.
Med (New York, N.Y.)
Verma S,Joshi CS,Silverstein RB,He M,Carter EB,Mysorekar IU
MA5-35946 was used in Immunohistochemistry to investigate the pathophysiology of SARS-CoV-2 at maternal-fetal interface in pregnant women who tested positive for the virus using RNA in situ hybridization (viral RNA), immunohistochemistry, and hematoxylin and eosin staining.
Fri May 14 00:00:00 EDT 2021