Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAASMARRLW PLLTRRGFRP RGGCISNDSP RRSFTTEKRN RNLLYEYARE GYSALPQLDI ERFCACPEEA AHALELRKGE LRSADLPAII STWQELRQLQ EQIRSLEEEK AAVTEAVRAL LANQDSGEVQ QDPKYQGLRA RGREIRKELV HLYPREAQLE EQFYLQALKL PNQTHPDVPV GDESQARVLH MVGDKPVFSF QPRGHLEIGE KLDIIRQKRL SHVSGHRSYY LRGAGALLQH GLVNFTFNKL LRRGFTPMTV PDLLRGAVFE GCGMTPNANP SQIYNIDPAR FKDLNLAGTA EVGLAGYFMD HTVAFRDLPV RMVCSSTCYR AETNTGQEPR GLYRVHHFTK VEMFGVTGPG LEQSSQLLEE FLSLQMEILT ELGLHFRVLD MPTQELGLPA YRKFDIEAWM PGRGRFGEVT SASNCTDFQS RRLHIMFQTE AGELQFAHTV NATACAVPRL LIALLESNQQ KDGSVLVPPA LQSYLGTDRI TAPTHVPLQY IGPNQPRKPG LPGQPAVS
This gene encodes the mitochondrial seryl-tRNA synthetase precursor, a member of the class II tRNA synthetase family. The mature enzyme catalyzes the ligation of Serine to tRNA(Ser) and participates in the biosynthesis of selenocysteinyl-tRNA(sec) in mitochondria. The enzyme contains an N-terminal tRNA binding domain and a core catalytic domain. It functions in a homodimeric form, which is stabilized by tRNA binding. This gene is regulated by a bidirectional promoter that also controls the expression of mitochondrial ribosomal protein S12. Both genes are within the critical interval for the autosomal dominant deafness locus DFNA4 and might be linked to this disease. Multiple transcript variants encoding different isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 6.1.1.11; mitochondrial seryl-tRNA synthetase; serine tRNA ligase 2, mitochondrial; Serine--tRNA ligase, mitochondrial; serine-tRNA ligase; serine-tRNA ligase, mitochondrial; SerRS; SerRSmt; Seryl-tRNA synthetase; seryl-tRNA synthetase, mitochondrial; Seryl-tRNA(Ser/Sec) synthetase
Gene Aliases: mtSerRS; SARS; SARS2; SARSM; SerRS; SerRSmt; SERS; SYS
UniProt ID: (Human) Q9NP81
Entrez Gene ID: (Human) 54938
Molecular Function:
aminoacyl-tRNA synthetase
translational protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support