Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat
Essential for gene silencing: has a role in the structure of silenced chromatin. Plays a role in the developing brain. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: charged amino acid rich leucine zipper 1 homolog; Charged amino acid-rich leucine zipper 1; CRL1; disrupter of silencing 10; Disrupter of silencing SAS10; Something about silencing protein 10; UTP3 homolog
Gene Aliases: CRL1; CRLZ1; SAS10; UTP3
UniProt ID: (Human) Q9NQZ2
Entrez Gene ID: (Human) 57050
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support