Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MARVAWGLLW LLLGSAGAQY EKYSFRGFPP EDLMPLAAAY GHALEQYEGE SWRESARYLE AALRLHRLLR DSEAFCHANC SGPAPAAKPD PDGGRADEWA CELRLFGRVL ERAACLRRCK RTLPAFQVPY PPRQLLRDFQ SRLPYQYLHY ALFKANRLEK AVAAAYTFLQ RNPKHELTAK YLNYYQGMLD VADESLTDLE AQPYEAVFLR AVKLYNSGDF RSSTEDMERA LSEYLAVFAR CLAGCEGAHE QVDFKDFYPA IADLFAESLQ CKVDCEANLT PNVGGYFVDK FVATMYHYLQ FAYYKLNDVR QAARSAASYM LFDPKDSVMQ QNLVYYRFHR ARWGLEEEDF QPREEAMLYH NQTAELRELL EFTHMYLQSD DEMELEETEP PLEPEDALSD AEFEGEGDYE EGMYADWWQE PDAKGDEAEA EPEPELA
This nucleolar protein was first characterized because it was an autoantigen in cases on interstitial cystitis. The protein, with a predicted molecular weight of 50 kDa, appears to be localized in the particulate compartment of the interphase nucleolus, with a distribution distinct from that of nucleolar protein B23. During mitosis it is associated with chromosomes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endoplasmic reticulum protein SC65; leprecan-like 4; Leprecan-like protein 4; nucleolar autoantigen (55kD); Nucleolar autoantigen No55; nucleolar autoantigen, 55kDa; Prolyl 3-hydroxylase family member 4; Synaptonemal complex protein SC65
Gene Aliases: LEPREL4; NO55; NOL55; P3H4; SC65
UniProt ID: (Human) Q92791
Entrez Gene ID: (Human) 10609
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support