Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: QPSRKLSPTE PKNYGSYSTQ ASAAAATAEL LKKQEELNRK AEELDRRERE LQHAALGGTA TRQNNWPPLP SFCPVQPCFF QDISMEIPQE FQKTVSTMYY
This gene product belongs to the SCAMP family of proteins which are secretory carrier membrane proteins. They function as carriers to the cell surface in post-golgi recycling pathways. Different family members are highly related products of distinct genes, and are usually expressed together. These findings suggest that the SCAMPs may function at the same site during vesicular transport rather than in separate pathways. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: propin 1; Secretory carrier membrane protein 3; Secretory carrier-associated membrane protein 3
Gene Aliases: C1orf3; PROPIN1; SCAMP3
UniProt ID: (Human) O14828
Entrez Gene ID: (Human) 10067
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support