Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MQAAALPEEI RWLLEDAEEF LAEGLRNENL SAVARDHRDH ILRGFQQIKA RYYWDFQPQG GDIGQDSSDD NHSGTLGLSL TSDAPFLSDY QDEGMEDIVK GAQELDNVIK QGYLEKKSKD HSFFGSEWQK RWCVVSRGLF YYYANEKSKQ PKGTFLIKGY GVRMAPHLRR DSKKESCFEL TSQDRRSYEF TATSPAEARD WVDQISFLLK DLSSLTIPYE EDEEEEEKEE TYDDIDGFDS PSCGSQCRPT ILPGSVGIKE PTEEKEEEDI YEVLPDEEHD LEEDESGTRR KGDYASYYQG LWDCHGDQPD ELSFQRGDLI RILSKEYNMY GWWVGELNSL VGIVPKEYLT TAFEVEER
This gene encodes a T cell adaptor protein, a class of intracellular molecules with modular domains capable of recruiting additional proteins but that exhibit no intrinsic enzymatic activity. The encoded protein contains a unique N-terminal region followed by a PH domain and C-terminal SH3 domain. Along with the adhesion and degranulation-promoting adaptor protein, the encoded protein plays a critical role in inside-out signaling by coupling T-cell antigen receptor stimulation to the activation of integrins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory sperm binding protein Li 81p; pp55; SKAP-55; src family associated phosphoprotein 1; Src family-associated phosphoprotein 1; Src kinase-associated phosphoprotein 1; Src kinase-associated phosphoprotein of 55 kDa
Gene Aliases: HEL-S-81p; SCAP1; SKAP1; SKAP55
UniProt ID: (Human) Q86WV1
Entrez Gene ID: (Human) 8631
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support