Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MASFQRSNSH DKVRRIVAEE GRTARNLIAW SVPLESKDDD GKPKCQTGGK SKRTIQGTHK TTKQSTAVDC KITSSTTGDK HFDKSPTKTR HPRKIDLRAR YWAFLFDNLR RAVDEIYVTC ESDQSVVECK EVLMMLDNYV RDFKALIDWI QLQEKLEKTD AQSRPTSLAW EVKKMSPGRH VIPSPSTDRI NVTSNARRSL NFGGSTGTVP APRLAPTGVS WADKVKAHHT GSTASSEITP AQSCPPMTVQ KASRKNERKD AEGWETVQRG RPIRSRSTAV MPKVSLATEA TRSKDDSDKE NVCLLPDESI QKGQFVGDGT SNTIESHPKD SLHSCDHPLA EKTQFTVSTL DDVKNSGSIR DNYVRTSEIS AVHIDTECVS VMLQAGTPPL QVNEEKFPAE KARIENEMDP SDISNSMAEV LAKKEELADR LEKANEEAIA SAIAEEEQLT REIEAEENND INIETDNDSD FSASMGSGSV SFCGMSMDWN DVLADYEARE SWRQNTSWGD IVEEEPARPP GHGIHMHEKL SSPSRKRTIA ESKKKHEEKQ MKAQQLREKL REEKTLKLQK LLEREKDVRK WKEELLDQRR RMMEEKLLHA EFKREVQLQA IVKKAQEEEA KVNEIAFINT LEAQNKRHDV LSKLKEYEQR LNELQEERQR RQEEKQARDE AVQERKRALE AERQARVEEL LMKRKEQEAR IEQQRQEKEK AREDAARERA RDREERLAAL TAAQQEAMEE LQKKIQLKHD ESIRRHMEQI EQRKEKAAEL SSGRHANTDY APKLTPYERK KQCSLCNVLI SSEVYLFSHV KGRKHQQAVR ENTSIQGREL SDEEVEHLSL KKYIIDIVVE STAPAEALKD GEERQKNKKK AKKIKARMNF RAKEYESLME TKNSGSDSPY KAKLQRLAKD LLKQVQVQDS GSWANNKVSA LDRTLGEITR ILEKENVADQ IAFQAAGGLT ALEHILQAVV PATNVNTVLR IPPKSLCNAI NVYNLTCNNC SENCSDVLFS NKITFLMDLL IHQLTVYVPD ENNTILGRNT NKQVFEGLTT GLLKVSAVVL GCLIANRPDG NCQPATPKIP TQEMKNKPSQ GDPFNNRVQD LISYVVNMGL IDKLCACFLS VQGPVDENPK MAIFLQHAAG LLHAMCTLCF AVTGRSYSIF DNNRQDPTGL TAALQATDLA GVLHMLYCVL FHGTILDPST ASPKENYTQN TIQVAIQSLR FFNSFAALHL PAFQSIVGAE GLSLAFRHMA SSLLGHCSQV SCESLLHEVI VCVGYFTVNH PDNQVIVQSG RHPTVLQKLC QLPFQYFSDP RLIKVLFPSL IAACYNNHQN KIILEQEMSC VLLATFIQDL AQTPGQAENQ PYQPKGKCLG SQDYLELANR FPQQAWEEAR QFFLKKEKK
CCNA2/CDK2 regulatory protein that transiently maintains CCNA2 in the cytoplasm.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: S phase cyclin A-associated protein in the endoplasmic reticulum; S phase cyclin A-associated protein in the ER; S-phase cyclin A-associated protein in the ER; Zinc finger protein 291
Gene Aliases: KIAA1454; MSTP063; SCAPER; Zfp291; ZNF291
UniProt ID: (Human) Q9BY12
Entrez Gene ID: (Human) 49855
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support