Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSASGVLSF TQQGWEQVLA KVKRAVVYLD AACAESLHWG CGSTRLLEAV GGPDCHLREF EPDAIGGGAK QPKAVFVLSC LLKGRTVEIL RDIICRSHFQ YCVVVTTVSH AVHLTANHVP AAAAAEMEGQ QPVFEQLEEK LCEWMGNMNY TAEVFHVPLL LAPVAPHFAL TPAFASLFPL LPQDVHLLNS ARPDKRKLGS LGDVDSTTLT PELLLQIRCL VSGLSSLCEH LGVREECFAV GSLSQVIAAD LANYAPAKNR KKTAAGRASV VFVDRTLDLT GAVGHHGDNL VEKIISALPQ LPGHTNDVMV NMIALTALHT EEENYNVVAP GCLSQSSDTT AKALWEALLN TK (1-351 aa encoded by BC032453)
SCFD2 may be involved in protein transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Neuronal Sec1; Sec1 family domain-containing protein 2; syntaxin binding protein 1-like 1; Syntaxin-binding protein 1-like 1
Gene Aliases: E430013M20Rik; SCFD2; STXBP1L1
UniProt ID: (Human) Q8WU76, (Mouse) Q8BTY8
Entrez Gene ID: (Human) 152579, (Mouse) 212986, (Rat) 498353
Molecular Function:
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support