Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Control:brain or gastrointestinal tract.
Human pancreas
Cellular Location: Cytoplasm. Secreted.
The encoded protein is a secreted calcium-binding protein which is found in the cytoplasm. It is related to calbindin D-28K and calretinin. This protein is thought to be involved in KCL-stimulated calcium flux and cell proliferation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Secretagogin
Gene Aliases: CALBL; DJ501N12.8; SCGN; SECRET; SEGN; setagin
UniProt ID: (Human) O76038
Entrez Gene ID: (Human) 10590
Molecular Function:
calmodulin-related
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support