Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This locus represents naturally occurring read-through transcription from the neighboring IQ motif containing J (IQCJ) and schwannomin interacting protein 1 (SCHIP1) genes. Alternative splicing results in multiple transcript variants that are composed of in-frame exons from each individual gene. The resulting fusion products are thought to be components of the multimolecular complexes of axon initial segments and nodes of Ranvier, and they may play a role in calcium-mediated responses.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: neurofibromatosis 2 interacting protein; SCHIP-1; schwannomin interacting protein 1; Schwannomin-interacting protein 1
Gene Aliases: Nf2ip; SCHIP-1; SCHIP1
UniProt ID: (Human) P0DPB3, (Mouse) P0DPB4
Entrez Gene ID: (Dog) 478678, (Human) 29970, (Mouse) 30953, (Rat) 295105
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support