Aliquoting is unnecessary for -20°C storage.
Selenocysteine lyase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hSCL; Selenocysteine lyase
Gene Aliases: hSCL; SCL; SCLY
UniProt ID: (Human) Q96I15, (Rat) Q68FT9
Entrez Gene ID: (Human) 51540, (Rat) 363285
Molecular Function:
lyase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support