Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SSAMQAPTV CIYLNKNGST GPHLDKKKVQ QLPDHFGPAR ASVVLQQAVQ ACIDCAYHQK TVFSFLKQGH GGEVISAVFD REQHTLNLPA VNSITYVLRF LEKLCHNLRS DNLFGNQPFT QTHLSLTAIE YSHSHDRYLP GETFVLGNSL ARSLEPHSDS MDSASNPTNL VSTSQRHRPL LSSCGLPPST ASAVRRLCSR GVLKGSNERR DMESFWKLNR SPGSDRYLES RDASRLSGRD PSSWTVEDVM QFVREADPQL GPHADLFRKH EIDGKALLLL RSDMMMKYMG LKLGPALKLS YHIDRLKQGK F (361-660 aa encoded by BC021252)
Associates with Polycomb group (PcG) multiprotein complexes; the complex class is required to maintain the transcriptionally repressive state of some genes. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Polycomb protein SCMH1; scmh1; sex comb on midleg 1; Sex comb on midleg homolog 1; sex comb on midleg-like 1; sex comb on midleg-like 3
Gene Aliases: AI315320; AI851618; SCMH1; Scml1; Scml3
UniProt ID: (Human) Q96GD3, (Mouse) Q8K214
Entrez Gene ID: (Human) 22955, (Mouse) 29871, (Rat) 362581
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support