Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: EKLCHNLRSD NLFGNQPFTQ THLSLTAIEY SHSHDRYLPG ETFVLGNSLA RSLEPHSDSM DSASNPTNLV STSQRHRPLL SSCGLPPSTA SAVRRLCSR
Associates with Polycomb group (PcG) multiprotein complexes; the complex class is required to maintain the transcriptionally repressive state of some genes. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Polycomb protein SCMH1; Sex comb on midleg homolog 1
Gene Aliases: SCMH1; Scml3
UniProt ID: (Human) Q96GD3
Entrez Gene ID: (Human) 22955
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support