Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total SCN3A.
Mediates the voltage-dependent sodium ion permeability of excitable membranes. Assuming opened or closed conformations in response to the voltage difference across the membrane, the protein forms a sodium-selective channel through which Na+ ions may pass in accordance with their electrochemical gradient.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brain III voltage-gated sodium channel; Sodium channel protein brain III subunit alpha; Sodium channel protein type 3 subunit alpha; Sodium channel protein type III subunit alpha; sodium channel, voltage gated, type III alpha subunit; sodium channel, voltage-gated, type III, alpha polypeptide; Voltage-gated sodium channel subtype III; Voltage-gated sodium channel subunit alpha Nav1.3
Gene Aliases: KIAA1356; NAC3; Nav1.3; SCN3A
UniProt ID: (Human) Q9NY46
Entrez Gene ID: (Human) 6328
Molecular Function:
voltage-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support