Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
Voltage-gated sodium channels are selective ion channels that regulate the permeability of sodium ions in excitable cells. During the propagation of an action potential, sodium channels allow an influx of sodium ions, which rapidly depolarize the cell. The three glycoproteins that comprise the voltagegated sodium channel proteins include a pore-forming alpha subunit, a noncovalently associated beta1 subunit and a disulfide-linked beta2 subunit. The two beta subunits regulate the level of channel expression, modulate gating and function as cell adhesion molecules for cellular aggregation and cytoskeleton interaction. The alpha subunits of sodium channels type I and III are predominantly expressed in neuronal cell bodies and proximal processes, while type II alpha subunits are more abundant along axons. The beta1 subunit of sodium channel type I is expressed in brain, skeletal and cardiac muscle. In the brain, beta1 and beta2 are highly expressed in Purkinje cells, and beta1 is also expressed in the pyramidal cells of the deep cerebellar nuclei. Impaired voltage-gated sodium channels lead to a number of diseases including myotonia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CTC-264K15.6; KIAA1356; MED; NAC1; NAC2; NAC3; Nav1.1; Nav1.2; Nav1.3; Nav1.5; Nav1.6; Nav1.7; Nav1.8; Nav1.9; NENA; SCN1; SCN10A; SCN11A; SCN12A; SCN1A; SCN2A; SCN2A1; SCN2A2; SCN3A; SCN5A; SCN8A; SCN9A; skeletal muscle sodium channel alpha subunit; skeletal muscle voltage-dependent sodium channel type IV alpha subunit; SkM1; SNS2; sodium channel alpha-subunit; Sodium channel protein skeletal muscle subunit alpha; Sodium channel protein type 4 subunit alpha; Sodium channel protein type IV subunit alpha; sodium channel, voltage gated, type IV alpha subunit; sodium channel, voltage-gated, type IV, alpha polypeptide; sodium channel, voltage-gated, type IV, alpha subunit; Voltage-gated sodium channel subunit alpha Nav1.4
Gene Aliases: CMS16; HOKPP2; HYKPP; HYPP; mH2; Na(V)1.4; NAC1A; Nav1.4; SCN4A; SkM1
UniProt ID: (Human) P35499, (Mouse) Q9ER60
Entrez Gene ID: (Human) 6329, (Mouse) 110880
Molecular Function:
voltage-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support