Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MSFKREGDDW SQLNVLKKRR VGDLLASYIP EDEALMLRDG RFACAICPHR PVLDTLAMLT AHRAGKKHLS SLQLFYGKKQ PGKERKQNPK HQNELRREET KAEAPLLTQT RLITQSALHR APHYNSCCRR KYRPEAPGPS VSLSPMPPSE VKLQSGKISR EPEPAAGPQA EESATVSAPA PMSPTRRRAL DHYLTLRSSG WIPDGRGRWV KDENVEFDSD EEEPPDLPLD
SCNM1 is a zinc finger protein and putative splicing factor. In mice, Scnm1 modifies phenotypic expression of Scn8a mutations.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Sodium channel modifier 1
Gene Aliases: SCNM1
UniProt ID: (Human) Q9BWG6
Entrez Gene ID: (Human) 79005
Molecular Function:
ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support